Learn More
Invitrogen™ FBXO32 Recombinant Rabbit Monoclonal Antibody (1F10Z5)

Description
Immunogen sequence: MPFLGQDWRS PGQNWVKTAD GWKRFLDEKS GSFVSDLSSY CNKEVYNKEN LFNSLNYDVA AKKRKKDMLN SKTKTQYFHQ EKWIYVHKGS TKERHGYCTL GEAFNRLDFS TAILDSRRFN.
Specifications
Specifications
| Antigen | FBXO32 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 1F10Z5 |
| Concentration | 1.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | FBXO32 |
| Gene Accession No. | Q91Z62, Q9CPU7 |
| Gene Alias | 4833442G10Rik; AI430017; atrogin 1; atrogin1; atrogin-1; atrophy gene 1; F-box only protein 32; F-box protein 32; Fbx32; fbxo25; FBXO32; FLJ32424; Gm20361; MAFbx; MGC108443; MGC137646; MGC33610; Muscle atrophy F-box protein; similar to F-box only protein 32 isoform 1; zgc:56479 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.