Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO33 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589740 Supplier Invitrogen™ Supplier No. PA589740
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VMHKSLDNMP NDEHWKALSR KSTSFRVYIM AFDIKSEDML KILKPSIPLE RIHFDSYITC VSGAIVDLIS RQYDKFLTHF ILMNDVIDTS GFPDLSDNRN EDPLVLLAWR CTKLSLLAIH GYTVWAHNLI AIARLRGSDL KVLEVTEESI DFDQGELADQ DVDPVHNLIE QVSLGLGQPW HAVMDIESLS VFTEPNRHFY REMQSFSEDI; Positive Samples: Rat brain, Rat testis.

FBXO33 is the substrate recognition component of a (SKP1-CUL1-F-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins. FBXO33 probably recognizes and binds to phosphorylated target proteins. Recognizes YBX1.Members of the F-box protein family, such as FBXO33, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, cullin, and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO33
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Fbxo33
Gene Alias 5730501N20Rik; AI642135; AIG30-12-1; BMND12; c14_5247; F-box only protein 33; F-box protein 33; FBX33; FBXO33
Gene Symbols Fbxo33
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 346-555 of human FBXO33 (NP_9760461).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 314157
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.