Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXO34 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP168997
Description
FBXO34 Polyclonal specifically detects FBXO34 in Rat samples. It is validated for Western Blot.Specifications
| FBXO34 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FBXO34 | |
| Synthetic peptides corresponding to Fbxo34 (F-box protein 34) The peptide sequence was selected from the N terminal of Fbxo34. Peptide sequence RDTLRTPMSHGKANGDVKARASYMKPTVLPSASLVKASSRKPFGILSPNV. | |
| Affinity purified | |
| RUO | |
| 55030 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| DKFZp547C162, F-box only protein 34, F-box protein 34, FBX34, FLJ20725, MGC126434, MGC126435, protein CGI-301 | |
| Rabbit | |
| 82 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: ; Pig: 86%; Rabbit: 86%; Guinea pig: 86%. | |
| Rat, Mouse, Rat, Bovine, Canine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction