Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO34 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562645
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562645 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562645 Supplier Invitrogen™ Supplier No. PA562645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ADEGKTKKGV LEAPDTQVNP VGSVSVDCGP SRADRCSPKE DQAWDGASQD CPPLPAGVSF HIDSAELEPG SQTAVKNSNR YDVE Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 51%.

Members of the F-box protein family, such as FBXO34, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO34
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Fbxo34
Gene Accession No. Q9NWN3
Gene Alias 2900057B08Rik; 5830426G16Rik; CGI-301; F-box only protein 34; F-box protein 34; Fbx34; FBXO34; LOW QUALITY PROTEIN: F-box only protein 34; protein CGI-301
Gene Symbols Fbxo34
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FBXO34. Recombinant protein control fragment (Product #RP-101531).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55030
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.