Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO36 Monoclonal Antibody (3D3)
GREENER_CHOICE

Catalog No. PIMA524517
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524517 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524517 Supplier Invitrogen™ Supplier No. MA524517
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: HLQGQTALIF GARILDYVIN FCKGKFDFLE RLSDDLLLTI ISYLDLEDIA RLCQTSHRFA KLCMSDKLWE QIVQSTCDTI TPDVRALAED TGWRQLFFTN.

Members of the F-box protein family, such as FBXO36, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO36
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene FBXO36
Gene Accession No. Q8NEA4
Gene Alias 0610008D19Rik; 1110020F21Rik; 2410002G19Rik; D1Ertd757e; F-box only protein 36; F-box protein 36; Fbx36; Fbxo36
Gene Symbols FBXO36
Host Species Mouse
Immunogen FBXO36 (NP_777559, 66 a.a. ~ 165 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130888
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.