Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO36 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565825
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565825 Supplier Invitrogen™ Supplier No. PA565825
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLSDDLLLTII SYLDLEDIAR LCQTSHRFAK LCMSDKLWEQ IVQSTCDTIT PDVRALAEDT GWRQLFFTNK LQLQRQLRKR KQ Highest antigen sequence identity to the following orthologs - mouse 76%, rat 80%.

Members of the F-box protein family, such as FBXO36, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO36
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FBXO36
Gene Accession No. Q8NEA4
Gene Alias 0610008D19Rik; 1110020F21Rik; 2410002G19Rik; D1Ertd757e; F-box only protein 36; F-box protein 36; Fbx36; Fbxo36
Gene Symbols FBXO36
Host Species Rabbit
Immunogen Recombinant Human FBXO36. Recombinant protein control fragment (Product #RP-104979).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130888
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.