Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO38 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501562446
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2446 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2446 Supplier Bethyl Laboratories, Inc Supplier No. A302378AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

Probably recognizes and binds to some phosphorylated proteins and promotes their ubiquitination and degradation. May coactivate KLF7, but does not seem to promote KLF7 ubiquitination.

Specifications

Antigen FBXO38
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene FBXO38
Gene Accession No. Q6PIJ6, Q8BMI0
Gene Alias Fbx38, FLJ13962, HMN2D, modulator of KLF7 activity homolog, MOKA, SP329
Gene Symbols FBXO38
Host Species Rabbit
Immunogen Between 825 and 875
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107035, 81545
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.