Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO38 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988837
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8837 Supplier Bioss Supplier No. BS9122R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Probably recognizes and binds to some phosphorylated proteins and promotes their ubiquitination and degradation. May coactivate KLF7, but does not seem to promote KLF7 ubiquitination.

Specifications

Antigen FBXO38
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FBXO38
Gene Accession No. Q6PIJ6
Gene Alias 6030410I24Rik; AU044865; AW214031; F-box only protein 38; F-box protein 38; F-box protein modulating KLF7 activity; Fbx38; FBXO38; FLJ13962; HMN2D; Modulator of KLF7 activity; modulator of KLF7 activity homolog; MoKA; SP329
Gene Symbols FBXO38
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FBXO38.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81545
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.