Learn More
FBXO4 Monoclonal Antibody (2F2)
FBXO4 Monoclonal Antibody (2F2)
Supplier:  Thermo Scientific MA521639
Catalog No. PIMA521639
Only null left
Add to Cart
Description
Description
FBXO4 Monoclonal Antibody for Western Blot, ICC/IF, IP, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FBXO4
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
Q9UKT5
FBXO4
FBXO4 (AAH48098, 1 a.a. ~ 387 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Immunoprecipitation, Immunocytochemistry, Western Blot
2F2
Unconjugated
FBXO4
1700096C12Rik; AI851261; AW494535; F-box only protein 4; F-box protein 4; F-box protein Fbx4; FBX4; Fbxo4
Mouse
Affinity Chromatography
RUO
26272
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.