Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FBXO4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564893
Description
Immunogen sequence: HEWQDEFSHIM AMTDPAFGSS GRPLLVLSCI SQGDVKRMPC FYLAHELHLN LLNHPWLVQD TEAETLTGFL NGIEWILEEV ESK Highest antigen sequence identity to the following orthologs - mouse 94%, rat 93%.
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternative splicing of this gene generates 2 transcript variants.
Specifications
FBXO4 | |
Polyclonal | |
Unconjugated | |
FBXO4 | |
1700096C12Rik; AI851261; AW494535; F-box only protein 4; F-box protein 4; F-box protein Fbx4; FBX4; Fbxo4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26272 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKT5 | |
FBXO4 | |
Recombinant Human FBXO4. Recombinant protein control fragment (Product #RP-105693). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction