Learn More
Invitrogen™ FBXO4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597311
Description
Immunogen sequence: DYMAVYRMCC PYTRRASKSS RPMYGAVTSF LHSLIIQNEP RFAMFGPGLE ELNTSLVLSL MSSEELCPTA GLPQRQIDGI GSGVNFQLNN QHKFNILILY STTRKERDRA REEHTSAVNK MFSRHNEGDD QQGSRYSVIP QIQKVCEVVD GFIYVANAEA HKRHEWQDEF SHIMAMTDPA FGSSGRPLLV LSCISQGDVK RMPCFYLAHE LHLNLLNHPW LVQDTEAETL TGFLNGIEWI LEEVESKRAR; Positive Samples: SH-SY5Y, Mouse spleen, Mouse kidney, Mouse liver; Cellular Location: Cytoplasm.
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternative splicing of this gene generates 2 transcript variants.
Specifications
FBXO4 | |
Polyclonal | |
Unconjugated | |
FBXO4 | |
1700096C12Rik; AI851261; AW494535; F-box only protein 4; F-box protein 4; F-box protein Fbx4; FBX4; Fbxo4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
106052, 26272 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.12 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CHQ0, Q9UKT5 | |
FBXO4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 138-387 of human FBXO4 (NP_0363081). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.