Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566167
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566167 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566167 Supplier Invitrogen™ Supplier No. PA566167
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ARREVFESTSF QGKEQAAGPS PAAPHLLHHH HHHAPLAHFP GDLVPASLPC EELAEPGLVP AAAARYALRE IEIPLGELFA RKSVAS Highest antigen sequence identity to the following orthologs - mouse 94%, rat 95%.

Substrate-recognition component of the SCF (SKP1-CUL1-F- box protein)-type E3 ubiquitin ligase complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO41
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FBXO41
Gene Accession No. Q8TF61
Gene Alias 9630017H13; D6Ertd538e; F-box only protein 41; F-box protein 41; FBX41; FBXO41; Kiaa1940; RGD1566130
Gene Symbols FBXO41
Host Species Rabbit
Immunogen Recombinant Human FBXO41. Recombinant protein control fragment (Product #RP-106851).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150726
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.