Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXO42 Antibody (OTI1H4), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271960F
Description
FBXO42 Monoclonal specifically detects FBXO42 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
FBXO42 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
FBXO42 | |
Recombinant protein fragment corresponding to amino acids 356-717 of human FBXO42 (NP_061867) produced in E.coli. | |
0.1 mL | |
54455.0 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry | |
OTI1H4 | |
Flow Cytometry, Immunohistochemistry | |
F-box only protein 42, F-box protein 42, Fbx42, JFK, Just one F-box and Kelch domain-containing protein, KIAA1332FBX42 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction