Learn More
Invitrogen™ FBXO42 Monoclonal Antibody (OTI2G7)

Description
FBXO42 Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | FBXO42 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2G7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | FBXO42 |
| Gene Accession No. | Q6P3S6 |
| Gene Alias | 6720460I06Rik; F-box only protein 42; F-box protein 42; Fbx42; Fbxo42; JFK; just one F-box and Kelch domain-containing protein; KIAA1332; LOC527807 protein; mKIAA1332 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.