Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO42 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA555224
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555224 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555224 Supplier Thermo Scientific Supplier No. PA555224

Rabbit Polyclonal Antibody

Immunogen sequence: HSLPPIARRL GHHPPQSLNV GKPLYQSMNC KPMQMYVLDI KDTKEKGRVK WKVFNSSSVV GPPETSLHTV VQGRGELIIF GGLMDKKQNV KY Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.

Members of the F-box protein family, such as FBXO42, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, cullin, and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO42
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FBXO42
Gene Accession No. Q6P3S6
Gene Alias 6720460I06Rik; F-box only protein 42; F-box protein 42; Fbx42; Fbxo42; JFK; just one F-box and Kelch domain-containing protein; KIAA1332; LOC527807 protein; mKIAA1332
Gene Symbols FBXO42
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FBXO42
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54455
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.