Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO43 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518188
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518188 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518188 Supplier Invitrogen™ Supplier No. PA518188
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, human and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Members of the F-box protein family, such as FBXO43, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1., cullin, and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO43
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene FBXO43
Gene Accession No. Q8CDI2
Gene Alias 4930533G20Rik; early mitotic inhibitor 2; EMI2; Endogenous meiotic inhibitor 2; ERP1; F-box only protein 43; F-box protein 43; FBX43; FBXO43; XErp1 homolog
Gene Symbols FBXO43
Host Species Goat
Immunogen Synthetic peptide sequence (PGSAQSKRNLKRL) corresponding to the C-terminus amino acids of FBXO43 (aa 628-640).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78803
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.