Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO43 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555197
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555197 Supplier Invitrogen™ Supplier No. PA555197
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ITHDFSDSSL CINDENACPE LLGSSVSGTT CGTDEDIFVT PISNLVANIR FNASQILSPS PEVRGSISTP EDSGFNSLSL EKSEDSLSDQ EGS Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 66%.

Members of the F-box protein family, such as FBXO43, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1., cullin, and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO43
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FBXO43
Gene Accession No. Q4G163
Gene Alias 4930533G20Rik; early mitotic inhibitor 2; EMI2; Endogenous meiotic inhibitor 2; ERP1; F-box only protein 43; F-box protein 43; FBX43; FBXO43; XErp1 homolog
Gene Symbols FBXO43
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FBXO43. Recombinant protein control fragment (Product #RP-93846).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286151
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.