Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXO44 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP224386SS
Description
FBXO44 Polyclonal specifically detects FBXO44 in Human samples. It is validated for Western Blot.Specifications
FBXO44 | |
Polyclonal | |
Western Blot 1:500-1:1000 | |
FBG3 DKFZp781J0852, F-box gene 3, F-box only protein 44, F-box protein 44, F-box protein FBX30, F-box/G-domain protein 3, FBX30FBX6AFbx44, FBX44, Fbxo6a, MGC14140 | |
Rabbit | |
Affinity Purified | |
RUO | |
93611 | |
Human | |
IgG |
Western Blot | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
FBXO44 | |
Reacts with residues 103-119 (EWKVEDLSRDQRKEFPN) of the human FBG3 protein. | |
0.025 mg | |
Primary | |
High cross reactivity: 94% identical to mouse and rat FBG3. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction