Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FBXO44 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582085
Description
Immunogen sequence: SLHRNLLHNP CAEEGFEFWS LDVNGGDEWK VEDLSRDQRK EFPNDQVRSQ ARLRVQVPAV RSAPVVRARA SGDLPARPGD HPAEERCQVE GGLPHILQLP.
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs family, similar to F-box only protein 2 and F-box only protein 6. Four alternatively spliced transcript variants encoding two distinct isoforms have been found for this gene.
Specifications
FBXO44 | |
Polyclonal | |
Unconjugated | |
FBXO44 | |
5730411K09; AV001623; DKFZp781J0852; F Box Gama 3; FBG3; F-box gene 3; F-box only protein 44; F-box only protein 6a; F-box protein 44; F-box protein FBX30; F-box/G-domain protein 3; FBX30; Fbx44; FBX6A; FBXO44; FBXO6A; MGC14140; RGD1562463; RP3-330O12.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
93611 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.20 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H4M3 | |
FBXO44 | |
Recombinant protein corresponding to Human FBXO44. Recombinant protein control fragment (Product #RP-102200). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction