Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO45 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982655
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2655 Supplier Bioss Supplier No. BS13150R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Component of E3 ubiquitin ligase complexes. Required for normal neuromuscular synaptogenesis, axon pathfinding and neuronal migration. Plays a role in the regulation of neurotransmission at mature neurons. May controls synaptic activity by controlling UNC13A via ubiquitin dependent pathway. Specifically recognizes TP73, promoting its ubiquitination and degradation.

Specifications

Antigen FBXO45
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FBXO45
Gene Accession No. P0C2W1
Gene Alias F-box only protein 45; F-box protein 45; F-box/SPRY domain-containing protein 1; FBX45; FBXO45; hFbxo45
Gene Symbols FBXO45
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FBXO45.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200933
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.