Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXO46 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548760
Description
Substrate-recognition component of the SCF (SKP1-CUL1-F- box protein)-type E3 ubiquitin ligase complex.
Specifications
FBXO46 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q6PJ61 | |
FBXO46 | |
KLH conjugated synthetic peptide between 584-612 amino acids from the C-terminal region of human FBXO46. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
FBXO46 | |
F-box only protein 46, F-box only protein 34-like, FBXO46, FBX46, FBXO34L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23403 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction