Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO5 Monoclonal Antibody (3D2D6)

FBXO5 Monoclonal Antibody (3D2D6)

Supplier:  LIFE TECHNOLOGIES 376600

Encompass_Preferred

Catalog No. 37-660-0


Only null left
Add to Cart

Description

Description

FBXO5 Monoclonal Antibody for Western Blot, ELISA

TRUSTED_SUSTAINABILITY
Specifications

Specifications

FBXO5
Monoclonal
0.5 mg/mL
PBS with 0.1% sodium azide
Q9UKT4
Fbxo5
Synthetic peptide derived from the C-terminal region of human Emi1 (early mitotic inhibitor 1, Fbx5) protein.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Western Blot
3D2D6
Unconjugated
Fbxo5
2510044I10Rik; C85305; Early mitotic inhibitor 1; Emi1; f-box only protein 31; F-box only protein 5; F-box protein 5; F-box protein Fbx5; FBX5; Fbxo31; FBXO5
Mouse
Protein A
RUO
26271
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.