Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583055
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583055 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583055 Supplier Invitrogen™ Supplier No. PA583055
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EENFGDSLQS CLLQIQSPDQ YPNKNLLPVL HFEKVVCSTL KKNAKRNPKV DREMLKEIIA RGNFRLQNII GRKMGLECVD ILSELFRRGL R Highest antigen sequence indentity to the following orthologs: Mouse - 77%, Rat - 72%.

F-box proteins, critical components of the conserved ubiquitin-protein ligase complex SCF (Skp1/Cdc53-Cullin1/F-box), recruit substrates for ubiquitination and consequent degradation through their specific protein-protein interaction domains. Novel human F-box proteins named FBG1, FBG2, FBG3, FBG4 and FBG5 constitute a subfamily of mammalian F-box proteins. FBG genes are expressed in a limited number of human tissues including kidney, liver, brain and muscle tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Fbxo5
Gene Accession No. Q9UKT4
Gene Alias 2510044I10Rik; C85305; Early mitotic inhibitor 1; Emi1; f-box only protein 31; F-box only protein 5; F-box protein 5; F-box protein Fbx5; FBX5; Fbxo31; FBXO5
Gene Symbols Fbxo5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FBXO5. Recombinant protein control fragment (Product #RP-94668).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26271
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.