Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116242
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116242 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116242 Supplier Thermo Scientific Supplier No. PA5116242
Add to Cart
Add to Cart

FBXO5 Polyclonal Antibody

FBXO5 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Fbxo5
Gene Accession No. Q9UKT4
Gene Alias 2510044I10Rik; C85305; Early mitotic inhibitor 1; Emi1; f-box only protein 31; F-box only protein 5; F-box protein 5; F-box protein Fbx5; FBX5; Fbxo31; FBXO5
Gene Symbols Fbxo5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 192-221 amino acids from the Central region of human FBXO5.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26271
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.