Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO5 Recombinant Rabbit Monoclonal Antibody (JG35-83)
GREENER_CHOICE

Catalog No. PIMA534738
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534738 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534738 Supplier Invitrogen™ Supplier No. MA534738
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: LOVO, Siha, Rat cerebellum tissue, human tonsil tissue, human colon cancer tissue, human kidney tissue, mouse cerebellum tissue.

F-box proteins, critical components of the conserved ubiquitin-protein ligase complex SCF (Skp1/Cdc53-Cullin1/F-box), recruit substrates for ubiquitination and consequent degradation through their specific protein-protein interaction domains. Novel human F-box proteins named FBG1, FBG2, FBG3, FBG4 and FBG5 constitute a subfamily of mammalian F-box proteins. FBG genes are expressed in a limited number of human tissues including kidney, liver, brain and muscle tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JG35-83
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Fbxo5
Gene Accession No. Q9UKT4
Gene Alias 2510044I10Rik; C85305; Early mitotic inhibitor 1; Emi1; f-box only protein 31; F-box only protein 5; F-box protein 5; F-box protein Fbx5; FBX5; Fbxo31; FBXO5
Gene Symbols Fbxo5
Host Species Rabbit
Immunogen Recombinant protein within Human Emi1 aa 128-244.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26271
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.