Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXO6 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298796PCP
Description
FBXO6 Polyclonal antibody specifically detects FBXO6 in Human samples. It is validated for Western BlotSpecifications
FBXO6 | |
Polyclonal | |
Western Blot | |
FBG2F-box/G-domain protein 2, F-box only protein 6, F-box protein 6, F-box protein Fbx6, F-box protein that recognizes sugar chains 2, Fbx6b, FBX6FBS2F-box protein FBG2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human FBXO6. | |
0.1 mL | |
Cancer, Cell Cycle and Replication | |
26270 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction