Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO6 Mouse anti-Human, Polyclonal, Abnova™

Catalog No. 89339179 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-179 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-179 Supplier Abnova Corporation Supplier No. H00026270B01P
Add to Cart
Add to Cart

Mouse Polyclonal Antibody.

FBXO6 Polyclonal antibody specifically detects FBXO6 in Human samples. It is validated for WB-Tr.

Specifications

Antigen FBXO6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description F-box protein 6
Gene FBXO6
Gene Accession No. NM_018438.4
Gene Alias FBG2, FBS2, FBX6, Fbx6b
Host Species Mouse
Immunogen FBXO6 (NP_060908.1, 1 a.a. ∼ 293 a.a) full-length human protein
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) 26270
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.