Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO6 Polyclonal Antibody

Catalog No. PIPA110884
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA110884 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA110884 Supplier Invitrogen™ Supplier No. PA110884
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-10884 detects FBG2 (F Box 2) in human, mouse,and rat samples. PA1-10884 has been successfully used in Western blot procedures, where a band of approximately 34 kDa is detected. PA1-10884 immunogen is a synthetic peptide corresponding to residues 268-284 of human FBG2.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class, and its C-terminal region is highly similar to that of rat NFB42 (neural F Box 42 kDa) which may be involved in the control of the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO6
Applications Western Blot
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene FBXO6
Gene Accession No. Q923V4, Q9NRD1, Q9QZN4
Gene Alias AA408845; FBG1; Fbg2; F-box only protein 6; F-box only protein 6 b; F-box only protein 6b; F-box protein 6; F-box protein FBG2; F-box protein Fbx6; F-box protein that recognizes sugar chains 2; F-box/G-domain protein 2; Fbs1; FBS2; FBX2; FBX6; Fbx6b; FBXO6; Fbxo6b; NFB42; RP3-330O12.3
Gene Symbols FBXO6
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues 268-284 of human FBG2.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192351, 26270, 50762
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.