Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXO6 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31533220UL
This item is not returnable.
View return policy
Description
FBXO6 Polyclonal antibody specifically detects FBXO6 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
FBXO6 | |
Polyclonal | |
PBS, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
FBG2F-box/G-domain protein 2, F-box only protein 6, F-box protein 6, F-box protein Fbx6, F-box protein that recognizes sugar chains 2, Fbx6b, FBX6FBS2F-box protein FBG2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 70-260 of human FBXO6 (NP_060908.1). | |
20 μg | |
Cancer, Cell Cycle and Replication | |
26270 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction