Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO7 Antibody, DyLight 680, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP297739FR

Catalog No. NB103199


Only null left
Add to Cart

Description

Description

FBXO7 Polyclonal specifically detects FBXO7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

FBXO7
Polyclonal
50 mM Sodium Borate
Rabbit
Antigen Affinity-purified
RUO
25793
Store at 4°C in the dark.
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 680
F-box only protein 7, F-box protein 7, FBX, FBX07, FBX7Fbx, PARK15DKFZp686B08113, PKPS
Produced in rabbits immunized with E. coli-derived Human FBXO7 fragment.
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.