Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FBXO7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542300
Description
Peptide sequence: PGETPSQFPP LRPRFDPVGP LPGPNPILPG RGGPNDRFPF RPSRGRPTDG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 83%; Horse: 92%; Human: 100%; Mouse: 92%; Pig: 93%; Rabbit: 100%; Rat: 93%; Zebrafish: 83%.
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and it may play a role in regulation of hematopoiesis. Alternatively spliced transcript variants of this gene have been identified with the full-length natures of only some variants being determined.
Specifications
FBXO7 | |
Polyclonal | |
Unconjugated | |
FBXO7 | |
2410015K21Rik; A230052G17Rik; AI117679; AW260462; C85118; DKFZp686B08113; F-box only protein 7; F-box protein 7; FBX; FBX07; FBX7; FBXO7; hypothetical protein LOC558097; PARK15; PKPS; si:ch211-286m4.2; Unknown (protein for MGC:134509); zgc:114096 | |
Rabbit | |
Affinity chromatography | |
RUO | |
25793 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y3I1 | |
FBXO7 | |
Synthetic peptide directed towards the middle region of human FBXO7. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction