Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXO8 Antibody (OTI3C5), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271794APC
Description
FBXO8 Monoclonal antibody specifically detects FBXO8 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
FBXO8 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
26269 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI3C5 | |
Western Blot, Immunohistochemistry | |
F-box only protein 8, F-box protein 8, F-box/SEC7 protein FBS, FBSFBX8F-box protein Fbx8 | |
Human recombinant protein fragment corresponding to amino acids 1-231 of human FBXO8(NP_036312) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction