Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO8 Monoclonal Antibody (OTI11F6)
GREENER_CHOICE

Catalog No. PIMA526889
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526889 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526889 Supplier Invitrogen™ Supplier No. MA526889
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FBXO8 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. It contains a C-terminal amino acid sequence that bears a significant similarity with a portion of yeast Sec7p, a critical regulator of vesicular protein transport. This human protein may interact with ADP-ribosylation factor(s)(ARFs) and exhibit ARF-GEF (guanine nucleotide exchange factor) activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI11F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Fbxo8
Gene Accession No. Q9NRD0
Gene Alias DC10; F-box only protein 8; F-box protein 8; F-box protein Fbx8; F-box/SEC7 protein FBS; FBS; Fbx8; FBXO8; UNQ1877/PRO4320
Gene Symbols Fbxo8
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-231 of FBXO8 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26269
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.