Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551763
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551763 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551763 Supplier Invitrogen™ Supplier No. PA551763
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WRVVRNQQLQ QEGYSEQGYL TREQSRRMAA SNISNTNHRK QVQGGIDIYH LLKARKSKEQ EGFINLEMLP PELSFTILSY LNATDLCLAS CVWQDLANDE LLWQGLCKST WGHCSIYNKN PPLGFSFRKL YMQ Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. It contains a C-terminal amino acid sequence that bears a significant similarity with a portion of yeast Sec7p, a critical regulator of vesicular protein transport. This human protein may interact with ADP-ribosylation factor(s)(ARFs) and exhibit ARF-GEF (guanine nucleotide exchange factor) activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO8
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Fbxo8
Gene Accession No. Q9NRD0
Gene Alias DC10; F-box only protein 8; F-box protein 8; F-box protein Fbx8; F-box/SEC7 protein FBS; FBS; Fbx8; FBXO8; UNQ1877/PRO4320
Gene Symbols Fbxo8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FBXO8. Recombinant protein control fragment (Product #RP-102198).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26269
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.