Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXO9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525475
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525475 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525475 Supplier Invitrogen™ Supplier No. PA525475
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FBXO9 Polyclonal Antibody for Western Blot

FBXO9 (F-box protein 9) is a Ubl conjugation pathway protein and it contains one F-box domain and one TPR repeat. FBXO9 may act as a potential sepsis biomarker or therapeutic target. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which functions in phosphorylation-dependent ubiquitination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXO9
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Fbxo9
Gene Accession No. Q8BK06, Q9UK97
Gene Alias 9030401P18Rik; AA986398; Cross-immune reaction antigen 1; dJ341E18.2; F-box only protein 9; f-box protein 9; F-box protein Fbx9; FBX9; Fbxo9; MNCb-2471; NY-REN-57; renal carcinoma antigen NY-REN-57; VCIA1
Gene Symbols Fbxo9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 346-373 amino acids from the C-terminal region of human FBXO9.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26268, 71538
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.