Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FBXO9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584633
Description
Immunogen sequence: PDIIWVFPPQ AEAEEDCHSD TVRADDDEEN ESPAETDLQA QLQMFRAQWM FELAPGVSSS NLENRPCRAA RGSLQKTSAD TKGKQE.
FBXO9 (F-box protein 9) is a Ubl conjugation pathway protein and it contains one F-box domain and one TPR repeat. FBXO9 may act as a potential sepsis biomarker or therapeutic target. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which functions in phosphorylation-dependent ubiquitination.
Specifications
FBXO9 | |
Polyclonal | |
Unconjugated | |
Fbxo9 | |
9030401P18Rik; AA986398; Cross-immune reaction antigen 1; dJ341E18.2; F-box only protein 9; f-box protein 9; F-box protein Fbx9; FBX9; Fbxo9; MNCb-2471; NY-REN-57; renal carcinoma antigen NY-REN-57; VCIA1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26268 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UK97 | |
Fbxo9 | |
Recombinant protein corresponding to Human FBXO9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction