Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXW10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543814
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543814 Supplier Invitrogen™ Supplier No. PA543814
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SPEKDHSSKS ATSQVYWTAK TQHTSLPLSK APENEHLLGA ASNPEEPWRN Sequence homology: Human: 100%.

Members of the F-box protein family, such as FBXW10, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains. Western blots using two different antibodies against two unique regions of this protein target confirm the same apparent molecular weight in our tests.Members of the F-box protein family, such as FBXW10, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXW10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FBXW10
Gene Accession No. Q5XX13
Gene Alias F-box and WD repeat domain containing 10; F-box and WD-40 domain protein 10; F-box and WD-40 domain-containing protein 10; F-box/WD repeat-containing protein 10; Fbw10; FBXW10; HREP; SM25H2; SM2SH2; testicular tissue protein Li 69; ubiquitin ligase specificity factor; Ubiquitin ligase-specificity factor
Gene Symbols FBXW10
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human FBXW10 (aa 180-229).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10517
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.