Learn More
Invitrogen™ FBXW11 Polyclonal Antibody

Description
Immunogen sequence: MEPDSVIEDK TIELMCSVPR Highest antigen sequence indentity to the following orthologs: Rat - 45%, Mouse - 100%.
Specifications
Specifications
| Antigen | FBXW11 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Fbxw11 |
| Gene Accession No. | Q9UKB1 |
| Gene Alias | 2310065A07Rik; AA536858; beta-transducin repeat-containing protein 2; BTRC2; BTRCP2; F-box and WD repeat domain containing 11; F-box and WD repeats protein beta-TrCP2; F-box and WD-40 domain protein 11; F-box and WD-40 domain protein 1B; F-box protein Fbw1b; F-box/WD repeat-containing protein 11; F-box/WD repeat-containing protein 1B; Fbw11; FBW1B; FBXW11; Fbxw1b; homologous to Slimb protein; HOS; hypothetical protein; KIAA0696 |
| Gene Symbols | Fbxw11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.