Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXW12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557972
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557972 Supplier Invitrogen™ Supplier No. PA557972
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PQVFLTESLL RPSEGSVPLS TFLPHKLCAS ACWTPKVKNR ITLMSQSSTG KKTEFITFDL TTKKTGGQTV IQAYEIASFQ VAAHLKCPIW MGA Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 43%.

Members of the F-box protein family, such as FBXW12, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603034), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXW12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FBXW12
Gene Accession No. Q6X9E4
Gene Alias F-box and WD repeat domain containing 12; F-box and WD-40 domain protein 12; F-box and WD-40 domain-containing protein 12; F-box- and WD40-repeat-containing protein; F-box only protein 35; F-box/WD repeat-containing protein 12; FBW12; FBXO12; FBXO35; FBXW12
Gene Symbols FBXW12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FBXW12. Recombinant protein control fragment (Product #RP-96423).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285231
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.