Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXW2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100525
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100525 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100525 Supplier Invitrogen™ Supplier No. PA5100525
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FBXW2.

F-box proteins are an expanding family of eukaryotic proteins characterized by an approximately 40 amino acid motif, the F box. Some F-box proteins have been shown to be critical for the ubiquitin-mediated degradation of cellular regulatory proteins. In fact, F-box proteins are one of the four subunits of ubiquitin protein ligases, called SCFs. SCF ligases bring ubiquitin conjugating enzymes to substrates that are specifically recruited by the different F-box proteins. Mammalian F-box proteins are classified into three groups based on the presence of either WD-40 repeats, leucine-rich repeats, or the presence or absence of other protein-protein interacting domains. This gene encodes the second identified member of the F-box gene family and contains multiple WD-40 repeats.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXW2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FBXW2
Gene Accession No. B2RZ17, Q60584, Q9UKT8
Gene Alias 2700071L08Rik; EMBL:AAI66990.1}; F-box and WD repeat domain containing 2; F-box and WD-40 domain protein 2; F-box and WD-40 domain-containing protein 2; F-box and WD-40 domain-containing protein 2 {ECO:0000312; F-box protein Fbw2; F-box/WD repeat-containing protein 2; FBW2; FBXW2; fbxw2 {ECO:0000312; FWD2; MD6; MGC117371; Protein MD6; RGD:1310460}
Gene Symbols FBXW2
Host Species Rabbit
Immunogen A synthesized peptide derived from human FBXW2(Accession Q9UKT8), corresponding to amino acid residues F382-T432.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26190, 30050, 311881
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.