Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FBXW2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518189
Description
This antibody is predicted to react with bovine, canine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
F-box proteins are an expanding family of eukaryotic proteins characterized by an approximately 40 amino acid motif, the F box. Some F-box proteins have been shown to be critical for the ubiquitin-mediated degradation of cellular regulatory proteins. In fact, F-box proteins are one of the four subunits of ubiquitin protein ligases, called SCFs. SCF ligases bring ubiquitin conjugating enzymes to substrates that are specifically recruited by the different F-box proteins. Mammalian F-box proteins are classified into three groups based on the presence of either WD-40 repeats, leucine-rich repeats, or the presence or absence of other protein-protein interacting domains. This gene encodes the second identified member of the F-box gene family and contains multiple WD-40 repeats.
Specifications
| FBXW2 | |
| Polyclonal | |
| Unconjugated | |
| FBXW2 | |
| 2700071L08Rik; EMBL:AAI66990.1}; F-box and WD repeat domain containing 2; F-box and WD-40 domain protein 2; F-box and WD-40 domain-containing protein 2; F-box and WD-40 domain-containing protein 2 {ECO:0000312; F-box protein Fbw2; F-box/WD repeat-containing protein 2; FBW2; FBXW2; fbxw2 {ECO:0000312; FWD2; MD6; MGC117371; Protein MD6; RGD:1310460} | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 26190, 30050, 311881 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| B2RZ17, Q60584, Q9UKT8 | |
| FBXW2 | |
| Synthetic peptide sequence (KRGSSFLAGEHPG) corresponding to the C-terminus amino acids of FBXW2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction