Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXW2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | FBXW2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
FBXW2 Polyclonal specifically detects FBXW2 in Human samples. It is validated for Western Blot.Specifications
FBXW2 | |
Polyclonal | |
Rabbit | |
Q9UKT8 | |
26190 | |
Synthetic peptides corresponding to FBXW2(F-box and WD repeat domain containing 2) The peptide sequence was selected from the middle region of FBXW2. Peptide sequence SLISRWPLPEYRKSKRGSSFLAGEASWLNGLDGHNDTGLVFATSMPDHSI. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
F-box and WD repeat domain containing 2, F-box and WD-40 domain protein 2, F-box and WD-40 domain-containing protein 2, F-box/WD repeat-containing protein 2, FBW2Fwd2, FWD2, Md6, MGC117371, Protein MD6 | |
FBXW2 | |
IgG | |
51 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title