Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXW4/SHFM3 Rabbit anti-Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc FBXW4401AP

Description
FBXW4/SHFM3 Polyclonal antibody specifically detects FBXW4/SHFM3 in Human, Mouse, Non-human Primate, Rat samples. It is validated for Dot Blot, ELISA, Immunoprecipitation, Western BlotSpecifications
FBXW4/SHFM3 | |
Polyclonal | |
Unconjugated | |
FBXW4 | |
Dac; dactylaplasia; dactylin; dactylyn; F-box and WD repeat domain containing 4; F-box and WD-40 domain protein 4; F-box and WD-40 domain-containing protein 4; F-box protein Fbw4; F-box/WD repeat protein 4; F-box/WD repeat-containing protein 4; FBW4; FBWD4; FBXW4; LOW QUALITY PROTEIN: F-box/WD repeat-containing protein 4; protein hagoromo; SHFM3; SHSF3 | |
Synthetic peptide taken within amino acid region 240-290 on human FBX4 protein. | |
200 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Dot Blot, ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
P57775, Q9JMJ2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
30838, 309444, 450689, 6468 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction