Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXW5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597208
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597208 Supplier Invitrogen™ Supplier No. PA597208
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRNKPYDVFG CWLTETSLIS GNLHRIGDIT SCSVLWLNNA FQDVESENVN VVKRLFKIQN LNASTVRTVM VADCSRFDSP DLLLEAGDPA TSPCRIFDLG SDNEEVVAGP APAHAKEGLR HFLDRVLEGR AQPQLSERML ETKVAELLAQ GHTKPPERSA TGAKSKYLIF TTGCLTYSPH QIGIKQILPH QMTTAGPVLG EGRGSDAFFD ALDHVIDIHG HIIGMGLSPD NRYLYVNSRA WPNGAVVADP MQPPPIAEEI DLLVFDLKTM REVRRALRAH RAYTPNDECF FIFLDVSRDF VASGAEDRHG YIWDRHYNIC LARLRHEDVV NSVVFSPQEQ ELLLTASDDA TIKAWRSPRT MRVLQAPRPR PRTFFSWLAS QRR Positive Samples: A-549; Cellular Location: Cytoplasm.

This gene encodes a member of the F-box protein family, members of which are characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into three classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene contains WD-40 domains, in addition to an F-box motif, so it belongs to the Fbw class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene, however, they were found to be nonsense-mediated mRNA decay (NMD) candidates, hence not represented.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXW5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FBXW5
Gene Accession No. Q969U6
Gene Alias AI159739; F-box and WD repeat domain containing 5; F-box and WD-40 domain protein 5; F-box and WD-40 domain-containing protein 5; F-box protein Fbw5; F-box/WD repeat-containing protein 5; FBW5; Fbxw5; PP3971; WD repeat-containing F-box protein FBW5
Gene Symbols FBXW5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 184-566 of human FBXW5 (NP_0618711).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54461
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.