Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXW7/Cdc4 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10088139AF405
Description
FBXW7/Cdc4 Polyclonal antibody specifically detects FBXW7/Cdc4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
FBXW7/Cdc4 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
AGOFBXO30, Archipelago homolog, archipelago, Drosophila, homolog of, CDC4FLJ16457, F-box and WD repeat domain containing 7, F-box and WD-40 domain protein 7 (archipelago homolog, Drosophila), F-box and WD-40 domain-containing protein 7, F-box protein FBW7, F-box protein FBX30, F-box protein SEL-10, F-box/WD repeat-containing protein 7, FBW7hCdc4, FBX30FLJ11071, FBXW6, hAgo, homolog of C elegans sel-10, SEL-10DKFZp686F23254, SEL10FBW6 | |
The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human F-box and WD-40 domain protein 7 using the numbering given in entry NP_361014.1 (GeneID 55294). | |
0.1 mL | |
Cancer | |
55294 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction