Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXW7 Monoclonal Antibody (OTI2E12), TrueMAB™, OriGene
SDP

Catalog No. 501679452
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-9452 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-9452 Supplier OriGene Supplier No. TA802858
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclin E, one of the activators of the cyclin-dependent kinase Cdk2, is critical for the initiation of DNA replication and other S-phase functions. The proper timing and amplitude of cyclin E expression seem to be important, because elevated levels of cyclin E have been associated with a variety of maligcies and constitutive expression of cyclin E leads to genomic instability. Phosphorylated cyclin E depends on the novel human gene, hCdc4, that was found to be mutated in a cell line derived from breast cancer that expressed extremely high levels of cyclin E.

Specifications

Antigen FBXW7
Applications Western Blot
Classification Monoclonal
Clone OTI2E12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FBXW7
Gene Accession No. Q969H0
Gene Alias AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4
Gene Symbols FBXW7
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-354 of human FBXW7 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55294
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.