Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FBXW7 Monoclonal Antibody (OTI4C11)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526563
Description
FBXW7 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
F-Box and WD Repeat Domain Containing 7 (FBXW7) is a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. FBXW7 was previously referred to as FBX30, and belongs to the Fbws class; in addition to an F-box, this protein contains 7 tandem WD40 repeats. This protein binds directly to cyclin E and probably targets cyclin E for ubiquitin-mediated degradation. Mutations in this gene are detected in ovarian and breast cancer cell lines, implicating the gene's potential role in the pathogenesis of human cancers. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
FBXW7 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q969H0 | |
FBXW7 | |
Human recombinant protein fragment corresponding to amino acids 1-354 of FBXW7 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI4C11 | |
Unconjugated | |
FBXW7 | |
1110001A17Rik; AGO; archipelago; Archipelago homolog; CDC4; DKFZp686F23254; F-box and WD repeat domain containing 7; F-box and WD repeat domain containing 7, E3 ubiquitin protein ligase; F-box and WD-40 domain protein 7; F-box and WD-40 domain protein 7 (archipelago homolog, Drosophila); F-box and WD-40 domain protein 7, archipelago homolog; F-box and WD-40 domain-containing protein 7; f-box only protein 30; F-box protein FBW7; F-box protein FBX30; F-box protein Fbxw6; F-box protein SEL-10; F-box/WD repeat-containing protein 7; F-box-WD40 repeat protein 6; FBW6; FBW7; Fbwd6; FBX30; Fbxo30; FBXW6; FBXW7; FLJ11071; FLJ16457; hAgo; hCdc4; homolog of C elegans sel-10; SEL10; SEL-10 | |
Mouse | |
Affinity chromatography | |
RUO | |
55294 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction