Learn More
FBXW7 Mouse anti-Human, Clone: OTI2E12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF802858

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cyclin E, one of the activators of the cyclin-dependent kinase Cdk2, is critical for the initiation of DNA replication and other S-phase functions. The proper timing and amplitude of cyclin E expression seem to be important, because elevated levels of cyclin E have been associated with a variety of maligcies and constitutive expression of cyclin E leads to genomic instability. Phosphorylated cyclin E depends on the novel human gene, hCdc4, that was found to be mutated in a cell line derived from breast cancer that expressed extremely high levels of cyclin E.Specifications
FBXW7 | |
Monoclonal | |
Unconjugated | |
FBXW7 | |
AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55294 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2E12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q969H0 | |
FBXW7 | |
Human recombit protein fragment corresponding to amino acids 1-354 of human FBXW7 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.