Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fbxw7 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8394R

Description
Cyclin E, one of the activators of the cyclin-dependent kinase Cdk2, is critical for the initiation of DNA replication and other S-phase functions. The proper timing and amplitude of cyclin E expression seem to be important, because elevated levels of cyclin E have been associated with a variety of malignancies and constitutive expression of cyclin E leads to genomic instability. Phosphorylated cyclin E depends on the novel human gene, hCdc4, that was found to be mutated in a cell line derived from breast cancer that expressed extremely high levels of cyclin E.Specifications
Fbxw7 | |
Polyclonal | |
Unconjugated | |
FBXW7 | |
1110001A17Rik; AGO; archipelago; Archipelago homolog; CDC4; DKFZp686F23254; F-box and WD repeat domain containing 7; F-box and WD repeat domain containing 7, E3 ubiquitin protein ligase; F-box and WD-40 domain protein 7; F-box and WD-40 domain protein 7 (archipelago homolog, Drosophila); F-box and WD-40 domain protein 7, archipelago homolog; F-box and WD-40 domain-containing protein 7; f-box only protein 30; F-box protein FBW7; F-box protein FBX30; F-box protein Fbxw6; F-box protein SEL-10; F-box/WD repeat-containing protein 7; F-box-WD40 repeat protein 6; FBW6; Fbw7; Fbwd6; FBX30; FBXO30; Fbxw6; Fbxw7; FLJ11071; FLJ16457; hAgo; hCdc4; homolog of C elegans sel-10; SEL10; SEL-10 | |
Rabbit | |
Protein A | |
RUO | |
55294 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q969H0 | |
FBXW7 | |
KLH conjugated synthetic peptide derived from human Fbxw7/hCDC4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction