Learn More
FBXW7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 552901AP150UL
Description
The FBXW7 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human FBXW7. This antibody recognizes human antigen. The FBXW7 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
FBXW7 | |
Polyclonal | |
Unconjugated | |
150 μL | |
55294 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
333 μg/mL | |
Rabbit | |
Primary | |
Human |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.